missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro[1,1-biphenyl]-4-carboxylic acid, 95%, Thermo Scientific™

Product Code. 10402304
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10402304 5 g 5g
10722133 250 mg 250mg
10551513 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10402304 Supplier Thermo Scientific Maybridge Supplier No. SEW05200DE
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 365-12-8
Molecular Formula C13H8FO2
Molecular Weight (g/mol) 215.20
MDL Number MFCD00278732
InChI Key SLKZDWAZOKIEEU-UHFFFAOYSA-M
Synonym 2'-fluorobiphenyl-4-carboxylic acid, 4-2-fluorophenyl benzoic acid, 2'-fluoro-1,1'-biphenyl-4-carboxylic acid, 2'-fluoro 1,1'-biphenyl-4-carboxylic acid, 2'-fluoro-biphenyl-4-carboxylic acid, maybridge3_006504, amtda094, acmc-1cr23, 4-2-fluorophenyl-benzoic acid, 2'-fluoro-4-biphenylcarboxylic acid
PubChem CID 715022
SMILES [O-]C(=O)C1=CC=C(C=C1)C1=CC=CC=C1F

Specifications

Assay Percent Range 95%
Packaging Amber Glass Bottle
Linear Formula FC6H4C6H4CO2H
Quantity 5 g
IUPAC Name 4-(2-fluorophenyl)benzoic acid
Formula Weight 216.21
Percent Purity 95%
Chemical Name or Material 2'-Fluoro[1, 1'-biphenyl]-4-carboxylic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.