missing translation for 'onlineSavingsMsg'
Learn More

2-Ethynylthiophene, ≥90%, Thermo Scientific™

Product Code. 10104053
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10104053 1 g Amber Glass Bottle 1g
10792984 250 mg Amber Glass Bottle 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10104053 Supplier Thermo Scientific Maybridge Supplier No. CC13047DA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 4298-52-6
Molecular Formula C6H4S
Molecular Weight (g/mol) 108.16
MDL Number MFCD03425129
InChI Key LWISLHRIEATKTM-UHFFFAOYSA-N
Synonym 2-ethynyl-thiophene, 2-thienylethine, 2-thiophenylacetylene, 2-thienyl acetylene, pubchem16123, thien-2-yl acetylene, thiophene, 2-ethynyl, thiophen-2-yl acetylene, 2-ethynylthiophene, acmc-209js9
PubChem CID 5246258
IUPAC Name 2-ethynylthiophene
SMILES C#CC1=CC=CS1

Specifications

CAS Min % 89.5
CAS Max % 100.0
Assay Percent Range 90%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 108.16
Percent Purity 90%
Chemical Name or Material 2-ethynylthiophene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.