missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Ethynylpyridine, TRC
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
Specifications
Specifications
| Chemical Name or Material | 2-Ethynylpyridine |
| CAS | 1945-84-2 |
| Molecular Formula | C7H5N |
| Quantity | 250 mg |
| Synonym | Pyridine, 2-ethynyl- (6CI, 7CI, 8CI, 9CI, ACI), 2-Ethynylpyridine (ACI), 2-Pyridinylethyne, 2-Pyridylacetylene, 2-Pyridylethyne, Pyridin-2-ylacetylene |
| InChI Formula | InChI=1S/C7H5N/c1-2-7-5-3-4-6-8-7/h1,3-6H |
| SMILES | C#Cc1ccccn1 |
| IUPAC Name | 2-ethynylpyridine |
| Molecular Weight (g/mol) | 103.12 |
| Formula Weight | 103.0422 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction