missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Ethylthiophene, 99%
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
Description
It is used as a heterocyclic building block. Also a maillard product used to provide a pleasant aroma to meat and coffee.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is used as a heterocyclic building block. Also a maillard product used to provide a pleasant aroma to meat and coffee.
Solubility
Not miscible or difficult to mix with water.
Notes
Store in cool, dry place in tightly closed container. Store away from oxidizing agent.
It is used as a heterocyclic building block. Also a maillard product used to provide a pleasant aroma to meat and coffee.
Solubility
Not miscible or difficult to mix with water.
Notes
Store in cool, dry place in tightly closed container. Store away from oxidizing agent.
Specifications
Specifications
| Density | 0.991 |
| Boiling Point | 133°C to 134°C |
| Flash Point | 21°C (69°F) |
| Odor | Stench |
| Refractive Index | 1.512 |
| Quantity | 100 g |
| UN Number | UN1993 |
| Beilstein | 105402 |
| Solubility Information | Not miscible or difficult to mix with water. |
| Formula Weight | 112.19 |
| Show More |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction