missing translation for 'onlineSavingsMsg'
Learn More

2-(ethylsulfanyl)ethane-1-thiol, TRC

Product Code. 30387225
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Product Code. Quantity
30387225 100 mg
30374497 50 mg
30381049 10 mg
3 options
This item is not returnable. View return policy

Product Code. 30387225

Brand: Toronto Research Chemicals TRCE902323100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 26750-44-7
Formula Weight 122.0224
InChI Formula InChI=1S/C4H10S2/c1-2-6-4-3-5/h5H,2-4H2,1H3
IUPAC Name 2-ethylsulfanylethanethiol
Molecular Formula C4H10S2
Molecular Weight (g/mol) 122.25
SMILES CCSCCS
Chemical Name or Material 2-(ethylsulfanyl)ethane-1-thiol

Specifications

Quantity 100 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.