missing translation for 'onlineSavingsMsg'
Learn More

2-Ethylpyrrole, 98%, Thermo Scientific Chemicals

Product Code. 10432841 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Glass bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10432841 1 g Glass bottle 1g
10702895 250 mg Glass bottle 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10432841 Supplier Thermo Scientific Acros Supplier No. 308770010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1551-06-0
Molecular Formula C6H9N
Molecular Weight (g/mol) 95.15
MDL Number MFCD00060477
InChI Key XRPDDDRNQJNHLQ-UHFFFAOYSA-N
PubChem CID 137075
IUPAC Name 2-ethyl-1H-pyrrole
SMILES CCC1=CC=CN1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Yellow to Brown
Density 0.9290g/mL
Boiling Point 142.0°C (15.0 mmHg)
Flash Point 62°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4985 to 1.5005
Quantity 1 g
Specific Gravity 0.929
Synonym 2-ethylpyrrole, 1h-pyrrole, 2-ethyl, pyrrole,2-ethyl, 1h-pyrrole,2-ethyl, acmc-209dbz, #, 2-ethylpyrrole, analytical standard, 2-ethylpyrrole, technical grade
Formula Weight 95.15
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 2-Ethylpyrrole
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.