missing translation for 'onlineSavingsMsg'
Learn More

2-Ethylimidazole, 99%, Thermo Scientific Chemicals

Catalog Number 10185080
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Glass bottle
Plastic Bottle
This item is not returnable. View return policy
1072-62-4
C5H8N2
96.13
MFCD00005192
PQAMFDRRWURCFQ-UHFFFAOYSA-N
2-ethylimidazole, 1h-imidazole, 2-ethyl, imidazole, 2-ethyl, 2-ethylimidazole 98+%, 2ez, 2-ethylimdazole, 2-ethyl imidazole, 2-ethyl-imidazole, pubchem15972
66130
2-ethyl-1H-imidazole
CCC1=NC=CN1
This item is not returnable. View return policy
1072-62-4
C5H8N2
96.13
MFCD00005192
PQAMFDRRWURCFQ-UHFFFAOYSA-N
2-ethylimidazole, 1h-imidazole, 2-ethyl, imidazole, 2-ethyl, 2-ethylimidazole 98+%, 2ez, 2-ethylimdazole, 2-ethyl imidazole, 2-ethyl-imidazole, pubchem15972
66130
2-ethyl-1H-imidazole
CCC1=NC=CN1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 98.5
CAS Max % 100.0
Melting Point 83.0°C to 87.0°C
Boiling Point 268.0°C
Flash Point 159°C
Assay Percent Range 1% max. Imidazole (GC)
Packaging Glass bottle
Quantity 100 g
Beilstein 23, 78
Solubility Information Solubility in water: 617g/L (20°C). Other solubilities: soluble in many organic solvents
Formula Weight 96.13
Percent Purity 99%
Chemical Name or Material 2-Ethylimidazole, 98%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 2-Ethylimidazole
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H318-Causes serious eye damage.
  • H315-Causes skin irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P362-Take off contaminated clothing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'