missing translation for 'onlineSavingsMsg'
Learn More

2-Ethylimidazole, 98%

Product Code. 11411097
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11411097 500 g 500g
11401097 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11411097 Supplier Thermo Scientific Alfa Aesar Supplier No. B24090.36
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1072-62-4 | C5H8N2 | 96.13 g/mol

2-Ethylimidazole is used as internal standard in the quantification of 4-(5-)methylimidazole (4MI) in coffee by GC-MS method. It is a versatile intermediate used as a building block for active ingredients as well as in epoxy curing.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Ethylimidazole is used as internal standard in the quantification of 4-(5-)methylimidazole (4MI) in coffee by GC-MS method. It is a versatile intermediate used as a building block for active ingredients as well as in epoxy curing.

Solubility
Soluble in water (617 g/L at 20°C)

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1072-62-4
Molecular Formula C5H8N2
Molecular Weight (g/mol) 96.13
MDL Number MFCD00005192
InChI Key PQAMFDRRWURCFQ-UHFFFAOYSA-N
Synonym 2-ethylimidazole, 1h-imidazole, 2-ethyl, imidazole, 2-ethyl, 2-ethylimidazole 98+%, 2ez, 2-ethylimdazole, 2-ethyl imidazole, 2-ethyl-imidazole, pubchem15972
PubChem CID 66130
IUPAC Name 2-ethyl-1H-imidazole
SMILES CCC1=NC=CN1

Specifications

Melting Point 78°C to 86°C
Boiling Point 268°C
Flash Point 159°C (318°F)
Quantity 500 g
Beilstein 106381
Solubility Information Soluble in water (617g/L at 20°C)
Formula Weight 96.13
Percent Purity 98%
Physical Form Crystals or Powder or Crystalline Powder or Flakes
Chemical Name or Material 2-Ethylimidazole

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.