missing translation for 'onlineSavingsMsg'
Learn More

2-Ethylhexylamine, 99%

Product Code. 13418498 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mL
250 mL
1 L
2.5 L
Packaging:
Glass Bottle
Unit Size:
1 L
100mL
2.5 L
250mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
13418498 2.5 L Glass Bottle 2.5 L
10678504 100 mL Glass Bottle 100mL
10378810 250 mL Glass Bottle 250mL
10195110 1 L Glass Bottle 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 13418498 Supplier Thermo Scientific Acros Supplier No. 271660025
 Restricted Item
Estimated Shipment: 25-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 104-75-6 | C8H19N | 129.25 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 104-75-6
Molecular Formula C8H19N
Molecular Weight (g/mol) 129.25
MDL Number MFCD00008148
InChI Key LTHNHFOGQMKPOV-UHFFFAOYSA-N
Synonym 2-ethylhexylamine, 1-hexanamine, 2-ethyl, 2-ethyl-1-hexylamine, 2-ethyl hexylamine, 2-ethylhexanamine, 1-amino-2-ethylhexane, hexylamine, 2-ethyl, beta-ethylhexylamine, 1-amino-2-ethylhexan, isooctylame
PubChem CID 7719
IUPAC Name 2-ethylhexan-1-amine
SMILES CCCCC(CC)CN

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point -76.0°C
Color Colorless
Density 0.7800g/mL
Boiling Point 169.0°C
Flash Point 50°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass Bottle
Linear Formula CH3(CH2)3CH(C2H5)CH2NH2
Refractive Index 1.4295 to 1.4315
Quantity 2.5 L
UN Number 2276
Beilstein 04,388
Specific Gravity 0.78
Solubility Information Solubility in water: 2.5g/L (20 c).
Viscosity 1.1 mPa.s (20°C)
Formula Weight 129.25
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 2-Ethylhexylamine
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • 2-Ethylhexylamine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H331-Toxic if inhaled.
  • H312-Harmful in contact with skin.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.