missing translation for 'onlineSavingsMsg'
Learn More

2-Ethylhexyl Stearate, TRC

Product Code. 30383654
Change view
Click to view available options
Quantity:
500 mg
1 g
5 g
Unit Size:
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30383654 500 mg 500mg
30376267 5 g -
30380809 1 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30383654 Supplier Toronto Research Chemicals Supplier No. TRCE918885500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 22047-49-0
Molecular Formula C26H52O2
Molecular Weight (g/mol) 396.68
Synonym Cetiol 868, Crodamol OS, Crodamol OS-LQ, Estol 1545, Exceparl EH-S, Exceparl ES-H, MB 876, Nikkol STO, Nissan Unister MB 876, Octyl Stearate, Pelemol OS, Stearic Acid 2-Ethylhexanyl Ester, Tegosoft OS, Unister MB 876, Wickenol 156, Ethyl Hexyl Stearate
IUPAC Name 2-ethylhexyl octadecanoate
SMILES CCCCCCCCCCCCCCCCCC(=O)OCC(CC)CCCC

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C26H52O2/c1-4-7-9-10-11-12-13-14-15-16-17-18-19-20-21-23-26(27)28-24-25(6-3)22-8-5-2/h25H,4-24H2,1-3H3
Formula Weight 396.3967
Chemical Name or Material 2-Ethylhexyl Stearate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.