missing translation for 'onlineSavingsMsg'
Learn More

2-Ethylhexyl Propanoate, TRC

Product Code. 30386808
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30386808 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30386808 Supplier Toronto Research Chemicals Supplier No. TRCE9188051G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-Ethylhexyl Propanoate
CAS 6293-37-4
Molecular Formula C11H22O2
Quantity 1 g
Synonym 2-Ethyl-1-hexanol propionate, 2-Ethylhexyl Propionate, NSC 8902, Propionic Acid 2-Ethylhexyl Ester
InChI Formula InChI=1S/C11H22O2/c1-4-7-8-10(5-2)9-13-11(12)6-3/h10H,4-9H2,1-3H3
SMILES O=C(CC)OCC(CC)CCCC
IUPAC Name 2-ethylhexyl propanoate
Molecular Weight (g/mol) 186.29
Formula Weight 186.162
Product Title
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.