missing translation for 'onlineSavingsMsg'
Learn More

2-Ethylhexyl methacrylate, 98%, stab. with 4-methoxyphenol

Product Code. 11344348
Change view
Click to view available options
Quantity:
250 mL
1 L
4 L
20 L
Unit Size:
1 L
20 L
250mL
4 L
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11344348 1 L 1 L
11364348 250 mL 250mL
11354348 4 L 4 L
11374348 20 L 20 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11344348 Supplier Thermo Scientific Alfa Aesar Supplier No. 042303.K2
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Monomer used in the preparation of polymethacrylate polymers | CAS: 688-84-6 | C12H22O2 | 198.306 g/mol

Monomer used in the preparation of polymethacrylate polymers

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 688-84-6
Molecular Formula C12H22O2
Molecular Weight (g/mol) 198.306
MDL Number MFCD00009494
InChI Key WDQMWEYDKDCEHT-UHFFFAOYSA-N
Synonym 2-ethylhexyl methacrylate, 2-ethyl-1-hexyl methacrylate, 2-ethylhexyl methacryate, 2-propenoic acid, 2-methyl-, 2-ethylhexyl ester, methacrylic acid 2-ethylhexyl ester, methacrylic acid, 2-ethylhexyl ester, methacrylate, 2-ethylisohexy, 2-ethylhexylmethacrylate, dsstox_cid_7293, dsstox_rid_78391
PubChem CID 12713
IUPAC Name 2-ethylhexyl 2-methylprop-2-enoate
SMILES CCCCC(CC)COC(=O)C(=C)C

Specifications

Melting Point -50°C
Color Colorless
Density 0.885 g/mL
Boiling Point 218°C
Flash Point 92°C (198°F)
Odor Ether-like
Refractive Index 1.438
Quantity 1 L
Beilstein 1769420
Formula Weight 198.31
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 2-Ethylhexyl methacrylate, Stabilized with 4-methoxyphenol
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.