missing translation for 'onlineSavingsMsg'
Learn More

2-Ethylhexyl Diphenyl Phosphate (90%), TRC

Product Code. 30369496
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30369496 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30369496 Supplier Toronto Research Chemicals Supplier No. TRCE9179801G
 Restricted Item
Estimated Shipment: 22-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-Ethylhexyl diphenyl phosphate
CAS 1241-94-7
Molecular Formula C20 H27 O4 P
Quantity 1 g
Synonym Phosphoric acid, 2-ethylhexyl diphenyl ester (6CI, 7CI, 8CI, 9CI, ACI), 2-Ethylhexyl diphenyl phosphate, Diphenyl 2-ethylhexyl phosphate, Disflamoll DPO, EHDPP, Octicizer, Phosflex 342, Phosflex 362, Santicizer 141, 2-Ethylhexyldiphenyl Phosphate
InChI Formula InChI=1S/C20H27O4P/c1-3-5-12-18(4-2)17-22-25(21,23-19-13-8-6-9-14-19)24-20-15-10-7-11-16-20/h6-11,13-16,18H,3-5,12,17H2,1-2H3
SMILES CCCCC(CC)COP(=O)(Oc1ccccc1)Oc2ccccc2
IUPAC Name 2-ethylhexyl diphenyl phosphate
Molecular Weight (g/mol) 362.4
Formula Weight 362.1647
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.