missing translation for 'onlineSavingsMsg'
Learn More

2-Ethylhexyl 2,3,4,5-Tetrabromobenzoate-d17, TRC

Product Code. 30382929
Change view
Click to view available options
Quantity:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30382929 1 mg
30376079 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30382929 Supplier Toronto Research Chemicals Supplier No. TRCE9187821MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1794752-19-4
Molecular Formula C15H1D17Br4O2
Molecular Weight (g/mol) 567.027
Synonym 2,3,4,5-Tetrabromobenzoic Acid 2-Ethylhexyl-d17 Ester, TBB-d17
IUPAC Name [1,1,2,3,3,4,4,5,5,6,6,6-dodecadeuterio-2-(1,1,2,2,2-pentadeuterioethyl)hexyl] 2,3,4,5-tetrabromobenzoate
SMILES CCCCC(CC)COC(=O)C1=CC(=C(C(=C1Br)Br)Br)Br

Specifications

Quantity 1 mg
InChI Formula InChI=1S/C15H18Br4O2/c1-3-5-6-9(4-2)8-21-15(20)10-7-11(16)13(18)14(19)12(10)17/h7,9H,3-6,8H2,1-2H3/i1D3,2D3,3D2,4D2,5D2,6D2,8D2,9D
Formula Weight 562.911
Chemical Name or Material 2-Ethylhexyl-2,3,4,5-tetrabromobenzoate D17
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.