missing translation for 'onlineSavingsMsg'
Learn More

2-Ethylhexane-1,3-diol, 97%

Product Code. p-7081070
missing translation for 'orderingAttributeHoverText'
Quantity:
10,000 mL
2500 mL
500 mL
missing translation for 'unitSize'
10000mL
2500mL
500mL
This item is not returnable. View return policy

Product Code. 11444263

missing translation for 'mfr': Thermo Scientific Alfa Aesar A10898.AP

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 94-96-2 | C8H18O2 | 146.23 g/mol

2-Ethylhexane-1,3-diol is used in organic synthesis and can be used to produce polyester resin, polyester plasticizers, polyurethane resin. It is also effective vermifuge to mosquitoes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Ethylhexane-1,3-diol is used in organic synthesis and can be used to produce polyester resin, polyester plasticizers, polyurethane resin. It is also effective vermifuge to mosquitoes.

Solubility
Soluble in water (42g/L at 20°C). Soluble in alcohol and ether.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. It is hygroscopic in nature.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 94-96-2
Molecular Formula C8H18O2
Molecular Weight (g/mol) 146.23
MDL Number MFCD00004578
InChI Key RWLALWYNXFYRGW-UHFFFAOYNA-N
Synonym 2-ethyl-1,3-hexanediol, ethohexadiol, 1,3-hexanediol, 2-ethyl, octylene glycol, ethyl hexanediol, carbide 6-12, repellent 612, rutgers 612, 6-12-insect repellent, diol-kyowa 8
PubChem CID 7211
ChEBI CHEBI:34273
IUPAC Name 2-ethylhexane-1,3-diol
SMILES CCCC(O)C(CC)CO

Specifications

Melting Point -40°C
Boiling Point 241°C to 249°C
Odor Odorless
Linear Formula CH3CH2CH2CHOHCH(CH2CH3)CH2OH
Quantity 500 mL
Solubility Information Soluble in water (42g/L at 20°C). Soluble in alcohol and ether.
Formula Weight 146.23
Percent Purity 97%
Chemical Name or Material 2-Ethylhexane-1,3-diol

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.