missing translation for 'onlineSavingsMsg'
Learn More

2-Ethylhexanal, 96%

Product Code. 11445467
Change view
Click to view available options
Quantity:
50 g
250 g
Unit Size:
250g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11445467 50 g 50g
11455467 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11445467 Supplier Thermo Scientific Alfa Aesar Supplier No. B23471.18
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 123-05-7 | C8H16O | 128.215 g/mol

2-Ethylhexanal is used as an intermediate for chemical synthesis and in the production of pharmaceuticals and odorous substances.  

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Ethylhexanal is used as an intermediate for chemical synthesis & in the production of disinfectants, pharmaceuticals and odorous substances.  

Solubility
Slightly soluble in water.

Notes
Air sensitive. Store in a cool, dry conditions, in well sealed conditions. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 123-05-7
Molecular Formula C8H16O
Molecular Weight (g/mol) 128.215
MDL Number MFCD00006987
InChI Key LGYNIFWIKSEESD-UHFFFAOYSA-N
Synonym hexanal, 2-ethyl, 2-ethylhexaldehyde, 2-ethylhexylaldehyde, 3-formylheptane, 2-ethylcaproaldehyde, butylethylacetaldehyde, ethylbutylacetaldehyde, ethylhexaldehyde, alpha-ethylcaproaldehyde, butyl ethyl acetaldehyde
PubChem CID 31241
IUPAC Name 2-ethylhexanal
SMILES CCCCC(CC)C=O

Specifications

Melting Point <-100°C
Density 0.819
Boiling Point 163°C
Flash Point 50°C (122°F)
Refractive Index 1.4155
Quantity 50 g
UN Number UN1989
Beilstein 1700556
Sensitivity Air Sensitive
Solubility Information Slightly soluble in water.
Formula Weight 128.21
Percent Purity 97%
Chemical Name or Material 2-Ethylhexanal
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.