missing translation for 'onlineSavingsMsg'
Learn More

2-(Ethylcarbamoyl)pyridine-5-boronic acid pinacol ester, 96%

Product Code. 15470826
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15470826 250 mg 250mg
15460826 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15470826 Supplier Thermo Scientific Alfa Aesar Supplier No. H54151.MD
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1006876-28-3 | C14H21BN2O3 | 276.143 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1006876-28-3
Molecular Formula C14H21BN2O3
Molecular Weight (g/mol) 276.143
MDL Number MFCD13191367
InChI Key TVMRHMZREIYXSR-UHFFFAOYSA-N
Synonym n-ethyl-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl picolinamide, 6-ethylcarbamoyl pyridine-3-boronic acid pinacol ester, 2-ethylcarbamoyl pyridine-5-boronic acid pinacol ester, 2-ethylaminocarbonyl pyridine-5-boronic acid pinacol ester, 2-n-ethylaminocarbonyl pyridine-5-boronic acid pinacol ester, 2-pyridinecarboxamide, n-ethyl-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, n-ethyl-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine-2-carboxamide, n-ethyl-5-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl picolinamide, 5-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl pyridine-2-carboxylic acid ethylamide, n-ethyl-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-2-pyridinecarboxamide
PubChem CID 59868917
IUPAC Name N-ethyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine-2-carboxamide
SMILES B1(OC(C(O1)(C)C)(C)C)C2=CN=C(C=C2)C(=O)NCC

Specifications

Melting Point 52°C to 54°C
Quantity 250 mg
Formula Weight 276.14
Percent Purity 96%
Chemical Name or Material 2-(Ethylcarbamoyl)pyridine-5-boronic acid pinacol ester

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.