missing translation for 'onlineSavingsMsg'
Learn More

2-Ethylbutyryl chloride, 99%

Product Code. 11430228
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11430228 25 g 25g
11440228 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11430228 Supplier Thermo Scientific Alfa Aesar Supplier No. B23774.14
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 2736-40-5 | C6H11ClO | 134.60 g/mol

2-Ethylbutyryl chloride was used in syntheses of bicyclic thiophene derivatives. It is used as an organic intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Ethylbutyryl chloride was used in syntheses of bicyclic thiophene derivatives. It is used as an organic intermediate.

Solubility
Reacts with water.

Notes
Moisture sensitive. Store under inert gas. Store away from oxidizing agents, amines, alcohols, active metals, water/ moisture.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2736-40-5
Molecular Formula C6H11ClO
Molecular Weight (g/mol) 134.60
MDL Number MFCD00018812
InChI Key SMUKODJVMQOSAB-UHFFFAOYSA-N
Synonym 2-ethylbutyryl chloride, 2-ethylbutyroyl chloride, butanoyl chloride, 2-ethyl, butyryl chloride, 2-ethyl, 2-ethylbutyrylchloride, diethylacetylchloride, diethylacetyl chloride, acmc-1cqal, 2-ethyl butyryl chloride, 2-ethyl-butyryl chloride
PubChem CID 75954
IUPAC Name 2-ethylbutanoyl chloride
SMILES CCC(CC)C(Cl)=O

Specifications

Density 0.98
Boiling Point 137°C to 139°C
Flash Point 33°C (91°F)
Odor Acrid
Refractive Index 1.425
Quantity 25 g
UN Number UN3094
Sensitivity Moisture sensitive
Solubility Information Reacts with water.
Formula Weight 134.61
Percent Purity 99%
Chemical Name or Material 2-Ethylbutyryl chloride
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.