missing translation for 'onlineSavingsMsg'
Learn More

2-Ethylbutyric Acid, TRC

Product Code. 30372433
Change view
Click to view available options
Quantity:
1 g
10 g
100 g
Unit Size:
1g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30372433 1 g 1g
30383644 10 g -
30380616 100 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30372433 Supplier Toronto Research Chemicals Supplier No. TRCE6784451G

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 88-09-5
Molecular Formula C6 H12 O2
Molecular Weight (g/mol) 116.16
Synonym 2-Ethylbutyric Acid, Butanoic acid, 2-ethyl- (9CI, ACI), 2-Ethylbutanoic acid (ACI), Butyric acid, 2-ethyl- (6CI, 7CI, 8CI), Butyric acid, α-ethyl- (3CI), 2-Ethyl-n-butyric acid, 2-Ethylbutyric acid, 3-Pentanecarboxylic acid, Acetic acid, diethyl-, Diethylacetic acid, NSC 11765, NSC 8758, α-Ethylbutyric acid
IUPAC Name 2-ethylbutanoic acid
SMILES CCC(CC)C(=O)O

Specifications

Quantity 1 g
InChI Formula InChI=1S/C6H12O2/c1-3-5(4-2)6(7)8/h5H,3-4H2,1-2H3,(H,7,8)
Formula Weight 116.0837
Chemical Name or Material 2-Ethylbutyric acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.