missing translation for 'onlineSavingsMsg'
Learn More

2-Ethylbenzoic acid, 97%

Product Code. p-4089506 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 10376624

Brand: Thermo Scientific Acros 378750010

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 612-19-1 | C9H10O2 | 150.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 612-19-1
Molecular Formula C9H10O2
Molecular Weight (g/mol) 150.18
MDL Number MFCD00045838
InChI Key CGMMPMYKMDITEA-UHFFFAOYSA-N
Synonym benzoic acid, ethyl, o-ethylbenzoic acid, 2-ethylbenzoicacidethylester, ethylbenzoic acid, pubchem9869, 2-ethyl-benzoic acid, benzoic acid,2-ethyl, benzoic acid, 2-ethyl, acmc-1ax4g, 2-ethylbenzoic acid
PubChem CID 34170
IUPAC Name 2-ethylbenzoic acid
SMILES CCC1=CC=CC=C1C(=O)O

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 62.0°C to 66.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Linear Formula CH3CH2C6H4COOH
Quantity 1 g
Formula Weight 150.18
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 2-Ethylbenzoic acid
Show More Show Less
compliance-icons
Product Identifier
  • 2-Ethylbenzoic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.