missing translation for 'onlineSavingsMsg'
Learn More

2-Ethylbenzaldehyde, 97%

Product Code. 15566006 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15566006 5 g 5g
15576006 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15566006 Supplier Thermo Scientific Acros Supplier No. 460550050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 22927-13-5 | C9H10O | 134.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 22927-13-5
Molecular Formula C9H10O
Molecular Weight (g/mol) 134.18
MDL Number MFCD02261766
InChI Key NTWBHJYRDKBGBR-UHFFFAOYSA-N
Synonym benzaldehyde, 2-ethyl, o-ethylbenzaldehyde, 2-ethyl benzaldehyde, 2-ethyl-benzaldehyde, ethylbenzaldehyde, 2-ethyl-benzaldehyd, 6-ethyl benzaldehyde, pubchem3074, acmc-20a49p
PubChem CID 123406
IUPAC Name 2-ethylbenzaldehyde
SMILES CCC1=CC=CC=C1C=O

Specifications

Density 1.0200g/mL
Boiling Point 209.0°C
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Refractive Index 1.538
Quantity 5 g
Specific Gravity 1.02
Formula Weight 134.18
Percent Purity 97%
Chemical Name or Material 2-Ethylbenzaldehyde
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.