missing translation for 'onlineSavingsMsg'
Learn More

2-Ethylanthraquinone 97%, ACROS Organics™

Product Code. 10438260 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
5g; Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10438260 5g; Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10438260 Supplier Thermo Scientific Acros Supplier No. 118070050

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 84-51-5
CAS Min % 96.0
CAS Max % 100.0
Melting Point 108°C to 111°C
Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Molecular Formula C16H12O2
MDL Number MFCD00001237
Quantity 5 g
Beilstein 07, I, 425
Synonym 2-ethylanthraquinone, 2-ethyl anthraquinone, 2-ethyl-9,10-anthraquinone, anthraquinone, 2-ethyl, 9,10-anthracenedione, 2-ethyl, usaf so-1, beta-ethylanthraquinone, unii-59yj81qzkd, 59yj81qzkd, 2-ethylanthra-9,10-quinone
InChI Key SJEBAWHUJDUKQK-UHFFFAOYSA-N
SMILES CCC1=CC2=C(C=C1)C(=O)C3=CC=CC=C3C2=O
IUPAC Name 2-ethylanthracene-9,10-dione
Molecular Weight (g/mol) 236.27
PubChem CID 6772
Formula Weight 236.27
Percent Purity ≥96.0%
Physical Form Fine Crystalline Powder
Chemical Name or Material 2-Ethylanthraquinone, 97%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.