missing translation for 'onlineSavingsMsg'
Learn More

2-Ethylaniline, TRC

Product Code. 30372303
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30372303 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30372303 Supplier Toronto Research Chemicals Supplier No. TRCE67854025G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-Ethylaniline
CAS 578-54-1
Molecular Formula C8 H11 N
Quantity 25 g
Synonym Aniline, o-ethyl- (8CI), 2-Ethylbenzenamine, 2-Ethylaniline, 2-Ethylphenylamine, NSC 62014, o-Aminoethylbenzene, o-Ethylaniline
InChI Formula InChI=1S/C8H11N/c1-2-7-5-3-4-6-8(7)9/h3-6H,2,9H2,1H3
SMILES CCc1ccccc1N
IUPAC Name 2-ethylaniline
Molecular Weight (g/mol) 121.1796
Formula Weight 121.0891
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.