missing translation for 'onlineSavingsMsg'
Learn More

2-Ethylaniline, 97%

Product Code. 11457197
Change view
Click to view available options
Quantity:
100 g
500 g
2500 g
Unit Size:
100g
2500g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11457197 100 g 100g
11467197 500 g 500g
11477197 2500 g 2500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11457197 Supplier Thermo Scientific Alfa Aesar Supplier No. A14984.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 578-54-1 | C8H11N | 121.183 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 578-54-1
Molecular Formula C8H11N
Molecular Weight (g/mol) 121.183
MDL Number MFCD00007751
InChI Key MLPVBIWIRCKMJV-UHFFFAOYSA-N
Synonym o-ethylaniline, benzenamine, 2-ethyl, aniline, o-ethyl, 2-ethylbenzenamine, o-aminoethylbenzene, aniline, 2-ethyl, 2-ethyl aniline, 2-ethylphenylamine, unii-gr557n47k6, ccris 2858
PubChem CID 11357
IUPAC Name 2-ethylaniline
SMILES CCC1=CC=CC=C1N

Specifications

Melting Point -47°C
Density 0.983
Boiling Point 208°C to 210°C
Flash Point 91°C (196°F)
Odor Characteristic
Refractive Index 1.56
Quantity 100 g
UN Number UN2273
Beilstein 1841268
Formula Weight 121.18
Percent Purity 97%
Chemical Name or Material 2-Ethylaniline
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.