missing translation for 'onlineSavingsMsg'
Learn More

2-Ethylacrolein, tech. 90%, stab. with 50ppm hydroquinone

Product Code. 11444887
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11444887 100 g 100g
11424887 5 g 5g
11434887 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11444887 Supplier Thermo Scientific Alfa Aesar Supplier No. B21839.22
 Restricted Item
Estimated Shipment: 18-09-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 922-63-4 | C5H8O | 84.12 g/mol

2-Ethylacrolein has been used in total synthesis of (±)-tabersonine, Aspidosperma alkaloid and synthesis of conjugate with bovine serum albumin tethered to Tn.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Ethylacrolein has been used in total synthesis of (+/-)-tabersonine, Aspidosperma alkaloid and synthesis of conjugate with bovine serum albumin tethered to Tn.

Solubility
Soluble in alcohol. Insoluble in water.

Notes
Air sensitive. Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store at 2-8°C.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 922-63-4
Molecular Formula C5H8O
Molecular Weight (g/mol) 84.12
MDL Number MFCD00010129
InChI Key GMLDCZYTIPCVMO-UHFFFAOYSA-N
Synonym 2-ethylacrolein, 2-methylenebutanal, 2-ethylpropenal, butanal, 2-methylene, ethacrolein, 2-ethylacrylaldehyde, 2-methylenebutyraldehyde, ethylacrolein, alpha-ethylacrolein, alpha-ethylacrylaldehyde
PubChem CID 70203
IUPAC Name 2-methylidenebutanal
SMILES CCC(=C)C=O

Specifications

Density 0.859
Boiling Point 92°C to 93°C
Flash Point 1°C (34°F)
Odor Pungent
Refractive Index 1.4245
Quantity 100 g
UN Number UN1992
Beilstein 1098378
Sensitivity Air sensitive
Solubility Information Soluble in alcohol. Insoluble in water.
Formula Weight 84.12
Percent Purity 90%
Grade Technical
Chemical Name or Material 2-Ethylacrolein, stabilized with 50ppm hydroquinone
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.