missing translation for 'onlineSavingsMsg'
Learn More

2-Ethyl N1,N3-Bis(descyclohexyl) N1,N3-Bis(trans-Cyclohexyl) Daprodustat 4-Dibenzoate, TRC

Product Code. 30387619
Change view
Click to view available options
Quantity:
2.5 mg
5 mg
10 mg
Unit Size:
10mg
2.5mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30387619 10 mg 10mg
30384421 2.5 mg 2.5mg
30370986 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30387619 Supplier Toronto Research Chemicals Supplier No. TRCE19337510MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-Ethyl N1,N3-Bis(descyclohexyl) N1,N3-Bis(trans-Cyclohexyl) Daprodustat 4-Dibenzoate
Analyte or Component Names 2-Ethyl N1,N3-Bis(descyclohexyl) N1,N3-Bis(trans-Cyclohexyl) Daprodustat 4-Dibenzoate
Shipping Condition Room Temperature
Molecular Formula C35 H39 N3 O10
Synonym (1R,1'R,4r,4'r)-(5-((2-Ethoxy-2-oxoethyl)carbamoyl)-6-hydroxy-2,4-dioxopyrimidine-1,3(2H,4H)-diyl)bis(cyclohexane-4,1-diyl) Dibenzoate
InChI Formula InChI=1S/C35H39N3O10/c1-2-46-28(39)21-36-30(40)29-31(41)37(24-13-17-26(18-14-24)47-33(43)22-9-5-3-6-10-22)35(45)38(32(29)42)25-15-19-27(20-16-25)48-34(44)23-11-7-4-8-12-23/h3-12,24-27,41H,2,13-21H2,1H3,(H,36,40)/t24-,25-,26-,27-
Solution Type Neat
SMILES CCOC(=O)CNC(=O)C1=C(O)N([C@@H]2CC[C@H](CC2)OC(=O)c3ccccc3)C(=O)N([C@@H]4CC[C@H](CC4)OC(=O)c5ccccc5)C1=O
IUPAC Name [4-[3-(4-benzoyloxycyclohexyl)-5-[(2-ethoxy-2-oxoethyl)carbamoyl]-6-hydroxy-2,4-dioxopyrimidin-1-yl]cyclohexyl] benzoate
Industry Type Pharmaceutical
Molecular Weight (g/mol) 661.698
Quantity 10 mg
Formula Weight 661.264
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.