missing translation for 'onlineSavingsMsg'
Learn More

2-Ethyl-6-methylaniline, 98%, Thermo Scientific Chemicals

Product Code. p-7020738
Click to view available options
Quantity:
500 g
2500 g
Unit Size:
2500g
500g
This item is not returnable. View return policy

Product Code. 11475153

Brand: Thermo Scientific Alfa Aesar A12796.36

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 24549-06-2
Molecular Formula C9H13N
Molecular Weight (g/mol) 135.21
MDL Number MFCD00007752
InChI Key JJVKJJNCIILLRP-UHFFFAOYSA-N
Synonym 6-ethyl-o-toluidine, 2-methyl-6-ethylaniline, benzenamine, 2-ethyl-6-methyl, o-toluidine, 6-ethyl, 6-ethyl-2-toluidine, 2-methyl-6-ethyl aniline, aniline, 2-methyl-6-ethyl, 6-ethyl-2-methylaniline, 2-ethyl-6-methyl-aniline, 2-ethyl-6-methylbenzenamine
PubChem CID 32485
IUPAC Name 2-ethyl-6-methylaniline
SMILES CCC1=CC=CC(=C1N)C

Specifications

Melting Point -25°C
Density 0.969
Boiling Point 226°C to 228°C
Flash Point 89°C (192°F)
Refractive Index 1.552
Quantity 500 g
Beilstein 2079468
Sensitivity Air and light sensitive
Formula Weight 135.21
Percent Purity 98%
Chemical Name or Material 2-Ethyl-6-methylaniline
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.