missing translation for 'onlineSavingsMsg'
Learn More

2-Ethyl-5,5-dimethyl-1,3-dioxane, 99%

Product Code. 15465067
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15465067 5 g 5g
15475067 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15465067 Supplier Thermo Scientific Alfa Aesar Supplier No. H32351.06
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 768-58-1 | C8H16O2 | 144.214 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 768-58-1
Molecular Formula C8H16O2
Molecular Weight (g/mol) 144.214
MDL Number MFCD16622268
InChI Key QSHOOPIYPOINNH-UHFFFAOYSA-N
Synonym solution in acetonitrile 1000mg/l, 1,3-dioxane, 2-ethyl-5,5-dimethyl
PubChem CID 69852
IUPAC Name 2-ethyl-5,5-dimethyl-1,3-dioxane
SMILES CCC1OCC(CO1)(C)C

Specifications

Density 0.88
Boiling Point 154°C
Flash Point 50°C (122°F)
Odor Characteristic
Quantity 5 g
UN Number UN1993
Formula Weight 144.21
Percent Purity 99%
Chemical Name or Material 2-Ethyl-5,5-dimethyl-1,3-dioxane

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.