missing translation for 'onlineSavingsMsg'
Learn More

2-Ethyl-4-methyl-1H-imidazole-5-carbaldehyde, 97%, Thermo Scientific™

Product Code. 10767485
Change view
Click to view available options
Quantity:
1 g
10 g
25 g
Unit Size:
10g
1g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10767485 25 g 25g
10603423 1 g 1g
10131132 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10767485 Supplier Thermo Scientific Maybridge Supplier No. SB01365EB

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 88634-80-4
Molecular Formula C7H10N2O
Molecular Weight (g/mol) 138.17
MDL Number MFCD00173727
InChI Key RLBMKAXUSJOVJI-UHFFFAOYSA-N
PubChem CID 2795884
IUPAC Name 2-ethyl-5-methyl-1H-imidazole-4-carbaldehyde
SMILES CCC1=NC(C=O)=C(C)N1

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Quantity 25 g
Synonym 2-ethyl-4-methyl-1h-imidazole-5-carbaldehyde, 2-ethyl-5-methyl-3h-imidazole-4-carbaldehyde, 1h-imidazole-5-carboxaldehyde,2-ethyl-4-methyl, 1h-imidazole-5-carboxaldehyde, 2-ethyl-4-methyl, 2-ethyl-5-formyl-4-methylimidazole, 2-ethyl-4-methylimidazole-5-carbaldehyde, 2-ethyl-4-methyl-1h-imidazole-5-carboxaldehyde, acmc-20lc80, 2-ethyl-4-methyl-5-formylimidazole, 1h-imidazole-4-carboxaldehyde, 2-ethyl-5-methyl
Formula Weight 138.17
Percent Purity 97%
Chemical Name or Material 2-Ethyl-4-methyl-1H-imidazole-5-carbaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.