missing translation for 'onlineSavingsMsg'
Learn More

2-Ethyl-4-methyl-1,3-dioxolane, cis + trans, 99%

Product Code. 15464877
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15464877 5 g 5g
15474877 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15464877 Supplier Thermo Scientific Alfa Aesar Supplier No. H32187.06
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 4359-46-0 | C6H12O2 | 116.16 g/mol

2-Ethyl-4-methyl-1,3-dioxolane, cis + trans, is used as a flavouring agent in food industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Ethyl-4-methyl-1,3-dioxolane, cis + trans, is used as a flavouring agent in food industry.

Solubility
Slightly soluble in water. Soluble in alcohol.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4359-46-0
Molecular Formula C6H12O2
Molecular Weight (g/mol) 116.16
MDL Number MFCD16622267
InChI Key CSZCLQLJVFLXLI-UHFFFAOYSA-N
Synonym 1,3-dioxolane, 2-ethyl-4-methyl, 1,3-dioxolane,2-ethyl-4-methyl, cis, 1,2-propanediol acetal, 1,3-dioxolane, 2-ethyl-4-methyl-, cis, cis-2-thyl-4-methyl-1,3-dioxolane, 2-ethyl-4-methyl-1,3-dioxolane, cis + trans, solution in acetonitrile 1000mg/l
PubChem CID 92210
IUPAC Name 2-ethyl-4-methyl-1,3-dioxolane
SMILES CCC1OCC(O1)C

Specifications

Density 0.904
Boiling Point 117°C
Flash Point 29°C (84°F)
Odor Characteristic
Refractive Index 1.403
Quantity 5 g
UN Number UN1993
Solubility Information Slightly soluble in water. Soluble in alcohol.
Formula Weight 116.16
Percent Purity 99%
Chemical Name or Material 2-Ethyl-4-methyl-1,3-dioxolane, Cis + Trans
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.