missing translation for 'onlineSavingsMsg'
Learn More

2-Ethyl-3-methylpyrazine, 98+%

Product Code. 11438963
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11438963 5 g 5g
11448963 25 g 25g
11458963 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11438963 Supplier Thermo Scientific Alfa Aesar Supplier No. A11092.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 15707-23-0 | C7H10N2 | 122.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15707-23-0
Molecular Formula C7H10N2
Molecular Weight (g/mol) 122.17
MDL Number MFCD00006150
InChI Key LNIMMWYNSBZESE-UHFFFAOYSA-N
Synonym pyrazine, 2-ethyl-3-methyl, 3-ethyl-2-methylpyrazine, 2-methyl-3-ethylpyrazine, 2-ethyl-3-methyl pyrazine, pyrazine, ethylmethyl, 2-ethyl-3-methyl-pyrazine, unii-9gf35mk66u, fema no. 3155, pyrazine, 3-ethyl-2-methyl, 2-ethyl-3-methylpyrazine, 9ci
PubChem CID 27457
IUPAC Name 2-ethyl-3-methylpyrazine
SMILES CCC1=NC=CN=C1C

Specifications

Density 0.987
Boiling Point 169°C to 171°C
Flash Point 57°C (134°F)
Odor Characteristic
Refractive Index 1.503
Quantity 5 g
UN Number UN1993
Beilstein 956775
Formula Weight 122.17
Percent Purity ≥98%
Chemical Name or Material 2-Ethyl-3-methylpyrazine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.