missing translation for 'onlineSavingsMsg'
Learn More

2-Ethyl-3-methoxypyrazine, 99%, Thermo Scientific™

Product Code. p-7022861
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11442488

Brand: Thermo Scientific Alfa Aesar A15817.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 25680-58-4
Molecular Formula C7H10N2O
Molecular Weight (g/mol) 138.17
MDL Number MFCD00006129
InChI Key DPCILIMHENXHQX-UHFFFAOYSA-N
Synonym 2-methoxy-3-ethylpyrazine, pyrazine, 2-ethyl-3-methoxy, 3-ethyl-2-methoxypyrazine, unii-zbm5ld0we9, 2-ethyl-3-methoxy-pyrazine, zbm5ld0we9, 2-methoxy-3-ethyl pyrazine, pyrazine, 3-ethyl-2-methoxy, acmc-20ai24, ksc494q2n
PubChem CID 33135
IUPAC Name 2-ethyl-3-methoxypyrazine
SMILES CCC1=NC=CN=C1OC

Specifications

Density 1.048
Boiling Point 90°C (40 mmHg)
Flash Point 69°C (156°F)
Refractive Index 1.502
Quantity 1 g
Formula Weight 138.17
Percent Purity 99%
Chemical Name or Material 2-Ethyl-3-methoxypyrazine
compliance-icons
Product Identifier
  • 2-Ethyl-3-methoxypyrazine
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : 2-ethyl-3-methoxy-pyrazine
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.