missing translation for 'onlineSavingsMsg'
Learn More

2-Ethyl-3,(5 or 6)-dimethylpyrazine, TRC

Product Code. 30385286
Change view
Click to view available options
Quantity:
50 g
100 g
250 g
Unit Size:
100g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30385286 50 g 50g
30386313 250 g 250g
30375797 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30385286 Supplier Toronto Research Chemicals Supplier No. TRCE91687050G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-Ethyl-3,6-dimethylpyrazine∣3,5-Dimethyl-2-ethylpyrazine
CAS 13360-65-1∣13925-07-0
Analyte or Component Names 2-Ethyl-3,6-dimethylpyrazine∣3,5-Dimethyl-2-ethylpyrazine
Shipping Condition Room Temperature
Molecular Formula C8H12N2∣C8H12N2
Synonym 3-ethyl-2,5-dimethylpyrazine, 2-ethyl-3,5-dimethylpyrazine
InChI Formula InChI=1S/C8H12N2/c1-4-8-7(3)9-5-6(2)10-8/h5H,4H2,1-3H3∣InChI=1S/C8H12N2/c1-4-8-7(3)10-6(2)5-9-8/h5H,4H2,1-3H3
Solution Type Neat
SMILES CCc1nc(C)cnc1C∣CCc1ncc(C)nc1C
IUPAC Name 3-ethyl-2,5-dimethylpyrazine∣2-ethyl-3,5-dimethylpyrazine
Industry Type Pharmaceutical
Molecular Weight (g/mol) 136.19∣136.19
Quantity 50 g
Formula Weight 136.1∣136.1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.