missing translation for 'onlineSavingsMsg'
Learn More

2-Ethyl-2-phenylmalonamide, TRC

Product Code. 30386502
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30386502 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30386502 Supplier Toronto Research Chemicals Supplier No. TRCE925455100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-Ethyl-2-phenylmalonamide
CAS 7206-76-0
Purity Grade Notes HPLC
Molecular Formula C11 H14 N2 O2
Quantity 100 mg
Synonym 2-Ethyl-2-phenylmalondiamide
InChI Formula InChI=1S/C11H14N2O2/c1-2-11(9(12)14,10(13)15)8-6-4-3-5-7-8/h3-7H,2H2,1H3,(H2,12,14)(H2,13,15)
SMILES CCC(C(=O)N)(C(=O)N)c1ccccc1
IUPAC Name 2-ethyl-2-phenylpropanediamide
Molecular Weight (g/mol) 206.24
Formula Weight 206.1055
Percent Purity >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.