missing translation for 'onlineSavingsMsg'
Learn More

2-Ethyl-2-phenyl-1,3-propanediol, TRC

Product Code. 30376884
Change view
Click to view available options
Quantity:
100 mg
250 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30376884 100 mg
30376285 250 mg
30374644 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30376884 Supplier Toronto Research Chemicals Supplier No. TRCE247650100MG
 Restricted Item
Estimated Shipment: 08-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 24765-56-8
Molecular Formula C11H16O2
Molecular Weight (g/mol) 180.24
IUPAC Name 2-ethyl-2-phenylpropane-1,3-diol
SMILES CCC(CO)(CO)c1ccccc1

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C11H16O2/c1-2-11(8-12,9-13)10-6-4-3-5-7-10/h3-7,12-13H,2,8-9H2,1H3
Formula Weight 180.115
Chemical Name or Material 2-Ethyl-2-phenyl-1,3-propanediol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.