missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ 2-Ethyl-2-oxazoline, 99%

Product Code. 11420897
Change view
Click to view available options
Quantity:
500 g
Unit Size:
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11420897 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.
Product Code. 11420897 Supplier Thermo Scientific™ Supplier No. B23287.36
ChemsChrom Mega Deals
Use promo code 27993
View all promotions
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.

CAS: 10431-98-8 | C5H9NO | 99.13 g/mol

2-ethyl-2-oxazoline is an organic intermediate and monomer that is used in water-soluble polymers, pharmaceuticals, cosmetics, specialty coatings and adhesives.

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 10431-98-8
Melting Point -62°
Density 0.981
Boiling Point 128-129°
Flash Point 29°(84°F)
Molecular Formula C5H9NO
MDL Number MFCD00005302
Quantity 500 g
UN Number UN2734
InChI Key NYEZZYQZRQDLEH-UHFFFAOYSA-N
SMILES CCC1=NCCO1
IUPAC Name 2-ethyl-4,5-dihydro-1,3-oxazole
Molecular Weight (g/mol) 99.13
Formula Weight 99.13
Percent Purity 99%
Physical Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.