missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Ethyl-2-methylsuccinic Acid, TRC
Specifications
Specifications
| Chemical Name or Material | 2-Ethyl-2-Methylsuccinic Acid |
| CAS | 631-31-2 |
| Molecular Formula | C7 H12 O4 |
| Quantity | 50 mg |
| Synonym | Butanedioic acid, 2-ethyl-2-methyl-, Succinic acid, 2-ethyl-2-methyl- (6CI,7CI,8CI), 2-Ethyl-2-methylsuccinic acid, 2-Methyl-2-ethylsuccinic acid, 3-Methyl-3-carboxyvaleric acid, NSC 38752, α-Methyl-α-ethylsuccinic acid, (2RS)-2-Ethyl-2-methylbutanedioic acid |
| InChI Formula | InChI=1S/C7H12O4/c1-3-7(2,6(10)11)4-5(8)9/h3-4H2,1-2H3,(H,8,9)(H,10,11) |
| SMILES | CCC(C)(CC(=O)O)C(=O)O |
| IUPAC Name | 2-ethyl-2-methylbutanedioic acid |
| Molecular Weight (g/mol) | 160.168 |
| Formula Weight | 160.074 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?