missing translation for 'onlineSavingsMsg'
Learn More

2-Ethyl-2-methyl-1,3-dioxolane, TRC

Product Code. 30369494
Change view
Click to view available options
Quantity:
1 g
10 g
50 g
Unit Size:
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30369494 50 g 50g
30371745 10 g -
30382963 1 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30369494 Supplier Toronto Research Chemicals Supplier No. TRCE92340050G

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 126-39-6
Molecular Formula C6H12O2
Molecular Weight (g/mol) 116.16
Synonym 2-Butanone Ethylene Ketal, 2-Butanone Cyclic 1,2-Ethanediyl Acetal, 2-Ethyl-2-methyldioxolane, 2-Methyl-2-ethyl-1,3-dioxolane, 2-Methyl-2-ethyldioxolane, NSC 829
IUPAC Name 2-ethyl-2-methyl-1,3-dioxolane
SMILES CCC1(C)OCCO1

Specifications

Quantity 50 g
InChI Formula InChI=1S/C6H12O2/c1-3-6(2)7-4-5-8-6/h3-5H2,1-2H3
Formula Weight 116.0837
Chemical Name or Material 2-Ethyl-2-methyl-1,3-dioxolane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.