missing translation for 'onlineSavingsMsg'
Learn More

2-Ethyl-2-(hydroxymethyl)-1,3-propanediol, 98%

Product Code. 11464404
Change view
Click to view available options
Quantity:
10,000 g
2500 g
500 g
Unit Size:
10000g
2500g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11464404 500 g 500g
11474404 2500 g 2500g
11484404 10,000 g 10000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11464404 Supplier Thermo Scientific Alfa Aesar Supplier No. A13084.36
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 77-99-6 | C6H14O3 | 134.18 g/mol

2-Ethyl-2-(hydroxymethyl)-1,3-propanediol acts as a precursor to alkyd resins, high-gloss coatings and ion exchange resins. It is also employed as a multifunctional monomer utilized for the production of coatings, ethoxylated and propoxylated trimethylolpropane derivatives. Further, it serves as a building block in the polymer industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Ethyl-2-(hydroxymethyl)-1,3-propanediol acts as a precursor to alkyd resins, high-gloss coatings and ion exchange resins. It is also employed as a multifunctional monomer utilized for the production of coatings, ethoxylated and propoxylated trimethylolpropane derivatives. Further, it serves as a building block in the polymer industry.

Solubility
Soluble in water.

Notes
Hygroscopic. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 77-99-6
Molecular Formula C6H14O3
Molecular Weight (g/mol) 134.18
MDL Number MFCD00004694
InChI Key ZJCCRDAZUWHFQH-UHFFFAOYSA-N
Synonym trimethylolpropane, 2-ethyl-2-hydroxymethyl propane-1,3-diol, ethriol, 2-ethyl-2-hydroxymethyl-1,3-propanediol, hexaglycerine, ettriol, etriol, trimethylol propane, tmp alcohol, 1,1,1-tris hydroxymethyl propane
PubChem CID 6510
IUPAC Name 2-ethyl-2-(hydroxymethyl)propane-1,3-diol
SMILES CCC(CO)(CO)CO

Specifications

Melting Point 59°C to 60°C
Boiling Point 296°C to 298°C
Flash Point 172°C (341°F)
Quantity 500 g
Beilstein 1698309
Solubility Information Soluble in water.
Formula Weight 134.18
Percent Purity 98%
Chemical Name or Material 2-Ethyl-2-(hydroxymethyl)-1,3-propanediol
compliance-icons
Signal Word
  • Warning
Hazard Category
  • Reproductive toxicity Category 2
Hazard Statement
  • H361fd-Suspected of damaging fertility. Suspected of damaging the unborn child.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.