missing translation for 'onlineSavingsMsg'
Learn More

2-Ethyl-2-(hydroxymethyl)-1,3-propanediol, 98%

Product Code. 10317440 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
500 g
1 kg
2.5 kg
10 kg
Packaging:
Plastic bottle
Plastic drum
Unit Size:
10kg
1kg
2.5kg
500g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10317440 500 g Plastic bottle 500g
10295970 1 kg Plastic bottle 1kg
10512381 2.5 kg Plastic bottle 2.5kg
10699833 10 kg Plastic drum 10kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10317440 Supplier Thermo Scientific Acros Supplier No. 164655000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 77-99-6 | C6H14O3 | 134.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 77-99-6
Molecular Formula C6H14O3
Molecular Weight (g/mol) 134.18
MDL Number MFCD00004694
InChI Key ZJCCRDAZUWHFQH-UHFFFAOYSA-N
Synonym trimethylolpropane, 2-ethyl-2-hydroxymethyl propane-1,3-diol, ethriol, 2-ethyl-2-hydroxymethyl-1,3-propanediol, hexaglycerine, ettriol, etriol, trimethylol propane, tmp alcohol, 1,1,1-tris hydroxymethyl propane
PubChem CID 6510
IUPAC Name 2-ethyl-2-(hydroxymethyl)propane-1,3-diol
SMILES CCC(CO)(CO)CO

Specifications

Acidity 0.002% max. (as formic acid)
CAS Min % 97.5
CAS Max % 100.0
Melting Point 56.0°C to 60.0°C
Color White
Boiling Point 160°C (0.7 hPa)
Flash Point 172°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Plastic bottle
Linear Formula C2H5C(CH2OH)3
Quantity 500 g
Beilstein 01, III, 2349
Solubility Information Solubility in water: soluble. Other solubilities: freely soluble in alcohols,insoluble in hydrocarbons
Viscosity 157 mPa.s (75°C)
Formula Weight 134.18
Percent Purity 98%
Physical Form Flakes
Chemical Name or Material 2-Ethyl-2-(hydroxymethyl)-1, 3-propanediol
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.