missing translation for 'onlineSavingsMsg'
Learn More

2-Ethyl-1H-indene, 97%

Product Code. 11486306
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11486306 1 g 1g
11496306 5 g 5g
11406316 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11486306 Supplier Thermo Scientific Alfa Aesar Supplier No. B23104.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 17059-50-6 | C11H12 | 144.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 17059-50-6
Molecular Formula C11H12
Molecular Weight (g/mol) 144.22
MDL Number MFCD01321239
InChI Key BSHJHVHMLRKHBZ-UHFFFAOYSA-N
PubChem CID 582328
IUPAC Name 2-ethyl-1H-indene
SMILES CCC1=CC2=CC=CC=C2C1

Spezifikation

Density 0.964
Boiling Point 94°C to 96°C (10 mmHg)
Flash Point 87°C (190°F)
Refractive Index 1.559
Quantity 1 g
Synonym 2-ethylindene, 2-ethyl-1-h-indene, 1h-indene, 2-ethyl, pubchem5396, 1h-indene,2-ethyl, acmc-20a5x3
Formula Weight 144.22
Percent Purity 97%
Chemical Name or Material 2-Ethyl-1H-indene

RUO – Research Use Only

Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.