missing translation for 'onlineSavingsMsg'
Learn More

2-Ethyl-1H-indene, 97%

Product Code. 11406316
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11406316 25 g 25g
11486306 1 g 1g
11496306 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11406316 Supplier Thermo Scientific Alfa Aesar Supplier No. B23104.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 17059-50-6 | C11H12 | 144.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 17059-50-6
Molecular Formula C11H12
Molecular Weight (g/mol) 144.22
MDL Number MFCD01321239
InChI Key BSHJHVHMLRKHBZ-UHFFFAOYSA-N
PubChem CID 582328
IUPAC Name 2-ethyl-1H-indene
SMILES CCC1=CC2=CC=CC=C2C1

Specifications

Density 0.964
Boiling Point 94°C to 96°C (10 mmHg)
Flash Point 87°C (190°F)
Refractive Index 1.559
Quantity 25 g
Synonym 2-ethylindene, 2-ethyl-1-h-indene, 1h-indene, 2-ethyl, pubchem5396, 1h-indene,2-ethyl, acmc-20a5x3
Formula Weight 144.22
Percent Purity 97%
Chemical Name or Material 2-Ethyl-1H-indene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.