missing translation for 'onlineSavingsMsg'
Learn More

2-Ethyl-1-butanol, 99%

Product Code. 11460107
Change view
Click to view available options
Quantity:
100 mL
500 mL
2500 mL
Unit Size:
100mL
2500mL
500mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11460107 100 mL 100mL
11470107 500 mL 500mL
11480107 2500 mL 2500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11460107 Supplier Thermo Scientific Alfa Aesar Supplier No. B21026.AE
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 97-95-0 | C6H14O | 102.18 g/mol

It is used as a perfuming agent in cosmetics industry. 2-Ethyl-1-butanol is used for the synthesis of various pharmaceutical compounds, such as novel branched Alkyl carbamates, acting as anticonvulsant agent. It is also a solvent used for various organic synthesis, separation processes and ionic liquids, and solvent mixtures for pharmaceutical applications . It is also used to make penetrating oils, corrosion inhibitors, plasticizers, and perfumes; as cleaning agent for printed circuits; and to improve flow of paints and varnishes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as a perfuming agent in cosmetics industry. 2-Ethyl-1-butanol is used for the synthesis of various pharmaceutical compounds, such as novel branched Alkyl carbamates, acting as anticonvulsant agent. It is also a solvent used for various organic synthesis, separation processes and ionic liquids, and solvent mixtures for pharmaceutical applications . It is also used to make penetrating oils, corrosion inhibitors, plasticizers, and perfumes; as cleaning agent for printed circuits; and to improve flow of paints and varnishes.

Solubility
Soluble in water (4 mg/ml at 25°C), alcohol, ether, and most organic solvents.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 97-95-0
Molecular Formula C6H14O
Molecular Weight (g/mol) 102.18
MDL Number MFCD00004744
InChI Key TZYRSLHNPKPEFV-UHFFFAOYSA-N
Synonym 2-ethyl-1-butanol, 1-butanol, 2-ethyl, 2-ethylbutanol, 2-ethylbutyl alcohol, pseudohexyl alcohol, 3-methylolpentane, 2-ethylbutanol-1, ethylbutanol, 3-pentylcarbinol, sec-pentylcarbinol
PubChem CID 7358
IUPAC Name 2-ethylbutan-1-ol
SMILES CCC(CC)CO

Specifications

Melting Point -52°C
Boiling Point 146°C to 148°C
Flash Point 58°C (136°F)
Quantity 100 mL
UN Number UN1987
Beilstein 1731254
Formula Weight 102.18
Percent Purity 99%
Chemical Name or Material 2-Ethyl-1-butanol
compliance-icons
Product Identifier
  • 2-Ethyl-1-butanol
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
Supplemental information
  • MIXTURE LIST-Contain: 2-Ethyl-1-butanol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.