missing translation for 'onlineSavingsMsg'
Learn More

2-Ethyl-1,3-hexanediol, 99%, mixture of isomers

Product Code. 10703671 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 mL
1 L
Packaging:
Glass bottle
Unit Size:
1 L
250mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10703671 250 mL Glass bottle 250mL
10783661 1 L Glass bottle 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10703671 Supplier Thermo Scientific Acros Supplier No. 118512500
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 94-96-2 | C8H18O2 | 146.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 94-96-2
Molecular Formula C8H18O2
Molecular Weight (g/mol) 146.23
MDL Number MFCD00004578
InChI Key RWLALWYNXFYRGW-UHFFFAOYNA-N
Synonym 2-ethyl-1,3-hexanediol, ethohexadiol, 1,3-hexanediol, 2-ethyl, octylene glycol, ethyl hexanediol, carbide 6-12, repellent 612, rutgers 612, 6-12-insect repellent, diol-kyowa 8
PubChem CID 7211
ChEBI CHEBI:34273
IUPAC Name 2-ethylhexane-1,3-diol
SMILES CCCC(O)C(CC)CO

Specifications

Acidity 0.05mg KOH/g max.
CAS Min % 98.5
CAS Max % 100.0
Melting Point -40°C
Density 0.9422g/mL
Boiling Point 243°C
Flash Point 136°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula CH3CH2CH2CH(OH)CH(C2H5)CH2OH
Refractive Index 1.4495 to 1.4525
Quantity 250 mL
Merck Index 15, 3797
Specific Gravity 0.9422
Solubility Information Solubility in water: 42g/L (20°C). Other solubilities: soluble in ethanol,isopropanol,propylene glycol,,castor oil and ether
Viscosity 323 mPa.s (20°C)
Formula Weight 146.23
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 2-Ethyl-1, 3-hexanediol, 99%
Show More Show Less
compliance-icons
Product Identifier
  • 2-Ethyl-1,3-hexanediol, mixture of isomers
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.