missing translation for 'onlineSavingsMsg'
Learn More

2-Ethoxypyridine-5-boronic acid pinacol ester, 99%, Thermo Scientific Chemicals

Product Code. 11404610
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11404610 1 g 1g
11494600 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11404610 Supplier Thermo Scientific Alfa Aesar Supplier No. H50137.03
 Restricted Item
Estimated Shipment: 21-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1072945-01-7 | Molecular Formula: C13H20BNO3 | Molecular Weight: 249.117 g/mol

2-Ethoxypyridine-5-boronic acid pinacol ester is used as raw material for synthesis of pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Ethoxypyridine-5-boronic acid pinacol ester is used as raw material for synthesis of pharmaceutical intermediates.

Solubility
Sparingly soluble in water (0.019 g/L) (25°C)

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1072945-01-7
Molecular Formula C13H20BNO3
Molecular Weight (g/mol) 249.117
MDL Number MFCD07781182
InChI Key GXUHWEZZHFBDFA-UHFFFAOYSA-N
Synonym 2-ethoxy-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, 6-ethoxypyridine-3-boronic acid pinacol ester, 2-ethoxy-5-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-pyridine, 2-ethoxypyridine-5-boronic acid pinacol ester, 2-ethoxypyridine-5-boronic acid, pinacol ester, pubchem20146, 6-ethoxypyridine-3-boronicacidpinacolester, 6-ethoxypyridin-3-ylboronic acid pinacol ester, 2-ethoxy-5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, 2-ethoxy-5-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl pyridine
PubChem CID 24208796
IUPAC Name 2-ethoxy-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
SMILES B1(OC(C(O1)(C)C)(C)C)C2=CN=C(C=C2)OCC

Specifications

Quantity 1 g
Solubility Information Sparingly soluble in water (0.019g/L) (25°C)
Formula Weight 249.12
Percent Purity 99%
Chemical Name or Material 2-Ethoxypyridine-5-boronic acid pinacol ester

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.