missing translation for 'onlineSavingsMsg'
Learn More

2-Ethoxypyridine-3-boronic acid pinacol ester, 95%, Thermo Scientific™

Product Code. 15244348 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
This item is not returnable. View return policy

Product Code. 15244348

Brand: Thermo Scientific Acros 454362500

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 848243-23-2
Molecular Formula C13H20BNO3
Molecular Weight (g/mol) 249.12
MDL Number MFCD07366721
InChI Key GRXNEZGBXVIORK-UHFFFAOYSA-N
Synonym 2-ethoxy-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, 2-ethoxy-3-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-pyridine, 2-ethoxypyridine-3-boronic acid pinacol ester, 2-ethoxypyridin-3-ylboronic acid pinacol ester, 2-ethoxy-3-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, pubchem16583, 2-ethoxypyridin-3-yl boronic acid pinacol ester
PubChem CID 17750191
IUPAC Name 2-ethoxy-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
SMILES CCOC1=C(C=CC=N1)B1OC(C)(C)C(C)(C)O1

Specifications

Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Quantity 250 mg
Formula Weight 249.12
Percent Purity 95%
Chemical Name or Material 2-Ethoxypyridine-3-boronic acid pinacol ester
compliance-icons
Product Identifier
  • 2-Ethoxypyridine-3-boronic acid pinacol ester, 95%
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.