missing translation for 'onlineSavingsMsg'
Learn More

2-Ethoxynicotinic acid, 98%, Thermo Scientific™

Product Code. 11489916
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11489916 10 g 10g
11499916 50 g 50g
11409926 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11489916 Supplier Thermo Scientific Alfa Aesar Supplier No. B20887.09
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 35969-54-1
Molecular Formula C8H9NO3
Molecular Weight (g/mol) 167.164
MDL Number MFCD00128788
InChI Key XCMJQQOMGWGGSI-UHFFFAOYSA-N
Synonym 2-ethoxynicotinic acid, 2-ethoxy-nicotinic acid, 3-pyridinecarboxylic acid, 2-ethoxy, 3-pyridinecarboxylicacid, 2-ethoxy, pubchem16563, acmc-1aft2, 2-ethoxy-3-pyridinecarboxylic acid, 2-ethoxypyridine-3-carboxylic acid
PubChem CID 2736345
IUPAC Name 2-ethoxypyridine-3-carboxylic acid
SMILES CCOC1=C(C=CC=N1)C(=O)O

Specifications

Quantity 10 g
Formula Weight 167.16
Percent Purity 98%
Chemical Name or Material 2-Ethoxynicotinic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.