missing translation for 'onlineSavingsMsg'
Learn More

2-Ethoxynaphthoic Acid, Mikromol™

Product Code. 30052219
Change view
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30052219 25 mg 25mg
30049575 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30052219 Supplier Mikromol Supplier No. MM0392.040025
 Restricted Item
Estimated Shipment: 30-09-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

2-Ethoxynaphthoic Acid
TRUSTED_SUSTAINABILITY

Especificaciones

Chemical Name or Material 2-Ethoxynaphthoic Acid
CAS 2224-00-2
Analyte or Component Names 2-Ethoxynaphthoic Acid
Shipping Condition Room Temperature
Molecular Formula C13 H12 O3
Certifications/Compliance ISO 17025
Synonym 2-Ethoxynaphthoic Acid
InChI Formula InChI=1S/C13H12O3/c1-2-16-11-8-7-9-5-3-4-6-10(9)12(11)13(14)15/h3-8H,2H2,1H3,(H,14,15)
SMILES CCOc1ccc2ccccc2c1C(=O)O
IUPAC Name 2-ethoxynaphthalene-1-carboxylic acid
Molecular Weight (g/mol) 216.23
Quantity 25 mg
Formula Weight 216.0786 g/mol
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.