missing translation for 'onlineSavingsMsg'
Learn More

2-Ethoxynaphthalene, 99%

Product Code. 11339629
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
This item is not returnable. View return policy

Product Code. 11339629

Brand: Thermo Scientific Alfa Aesar B23327.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 93-18-5 | C12H12O | 172.227 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 93-18-5
Molecular Formula C12H12O
Molecular Weight (g/mol) 172.227
MDL Number MFCD00016808
InChI Key GUMOJENFFHZAFP-UHFFFAOYSA-N
Synonym bromelia, naphthalene, 2-ethoxy, neroline, nerolin new, nerolin ii, ethyl 2-naphthyl ether, 2-naphthol ethyl ether, bromelia compound, beta-naphthyl ethyl ether, ethyl beta-naphthyl ether
PubChem CID 7129
IUPAC Name 2-ethoxynaphthalene
SMILES CCOC1=CC2=CC=CC=C2C=C1

Specifications

Melting Point 35°C to 37°C
Boiling Point 282°C
Flash Point 134°C (273°F)
Quantity 25 g
Beilstein 2042397
Merck Index 14,3752
Formula Weight 172.23
Percent Purity 99%
Chemical Name or Material 2-Ethoxynaphthalene

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.