missing translation for 'onlineSavingsMsg'
Learn More

(2-ethoxyethyl)hydrazine xHCl salt, TRC

Product Code. 30370435
Change view
Click to view available options
Quantity:
100 mg
500 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30370435 100 mg
30375276 1 g
30384498 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30370435 Supplier Toronto Research Chemicals Supplier No. TRCE677283100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 809282-62-0
Molecular Formula C4H12N2O . x(HCl)
Molecular Weight (g/mol) 177.07
Synonym Hydrazine, (2-ethoxyethyl)-, xHCl salt (9CI)
SMILES Cl.Cl.CCOCCNN

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C4H12N2O.2ClH/c1-2-7-4-3-6-5;;/h6H,2-5H2,1H3;2*1H
Formula Weight 176.0483
Chemical Name or Material (2-ethoxyethyl)hydrazine xHCl salt
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.